siRNA Details
| Gene Symbol: Gm12830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033617.1 | 4 | 1631 | |||
| GenBank mRNA | AK038383.1 | - | 4 | 1632 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12830| Gene Symbol: Gm12830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033617.1 | 4 | 1631 | |||
| GenBank mRNA | AK038383.1 | - | 4 | 1632 | ||