siRNA Details
| Gene Symbol: Gm12830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033617.1 | 3 | 449 | |||
| GenBank mRNA | AK038383.1 | - | 3 | 450 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB588621.2 | - | 3 | 450 | Chr. 4 - 114821720 - 114854492 [+] on Build GRCm38 | |

Gm12830
Genomic Map