siRNA Details
| Gene Symbol: Gm16036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139684.1 | - | 545 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY725574.1 | - | 4 | 541 | Chr. 5 - 141596425 - 141615999 [-] on Build GRCm38 | |

Gm16036
Genomic Map