siRNA Details
| Gene Symbol: Gm17745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038014.1 | 3 | 775 | |||
| GenBank mRNA | AK144329.1 | - | 3 | 776 | ||
| Gene Symbol: Gm3546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI450451.1 | - | 305 | |||

Gm17745
Genomic Map