siRNA Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK148950.1 | - | 1 | 1677 | ||
| AK149008.1 | - | 1 | 1525 | |||
| AK149239.1 | - | 1 | 1589 | |||
| Gene Symbol: 9430041P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AV162270.2 | - | 1 | 989 | ||
| BB093294.2 | - | 1 | 632 | |||

Dpysl3
Genomic Map