siRNA Details
| Gene Symbol: Ubap2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086314.1 | - | 6 | 2289 | ||
| AK164224.1 | - | 1424 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubap2l| Gene Symbol: Ubap2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086314.1 | - | 6 | 2289 | ||
| AK164224.1 | - | 1424 | ||||