siRNA Details
| Gene Symbol: 9030404E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045878.1 | 4 | 727 | |||
| GenBank mRNA | AK018484.1 | - | 4 | 728 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720657.1 | - | 4 | 728 | Chr. 16 - 30022137 - 30039479 [-] on Build GRCm38 | |

9030404E10Rik
Genomic Map