siRNA Details
| Gene Symbol: Gm30606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039837.1 | - | 3 | 673 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB199732.2 | - | 3 | 529 | Chr. 8 - 93707015 - 93719814 [-] on Build GRCm38 | |

Gm30606
Genomic Map