siRNA Details
| Gene Symbol: Gm11738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039839.1 | - | 1 | 143 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB634410.1 | - | 1 | 143 | Chr. 11 - 118114855 - 118116795 [+] on Build GRCm38 | |

Gm11738
Genomic Map