siRNA Details
| Gene Symbol: Gm11738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039839.1 | - | 2 | 983 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11738| Gene Symbol: Gm11738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039839.1 | - | 2 | 983 | ||