siRNA Details
Gene Symbol: C130046K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102388.1 | 3 | 2658 | |||
NR_102389.1 | 3 | 2647 | ||||
GenBank mRNA | AK142001.1 | - | 3 | 2642 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C130046K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102388.1 | 3 | 2658 | |||
NR_102389.1 | 3 | 2647 | ||||
GenBank mRNA | AK142001.1 | - | 3 | 2642 |