siRNA Details
| Gene Symbol: C130046K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102388.1 | 3 | 1082 | |||
| NR_102389.1 | 3 | 1071 | ||||
| GenBank mRNA | AK142001.1 | - | 3 | 1066 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C130046K22Rik| Gene Symbol: C130046K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102388.1 | 3 | 1082 | |||
| NR_102389.1 | 3 | 1071 | ||||
| GenBank mRNA | AK142001.1 | - | 3 | 1066 | ||