siRNA Details
| Gene Symbol: 1700001G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038077.1 | 1 | 312 | |||
| GenBank mRNA | AK005591.1 | - | 1 | 160 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001G11Rik| Gene Symbol: 1700001G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038077.1 | 1 | 312 | |||
| GenBank mRNA | AK005591.1 | - | 1 | 160 | ||