siRNA Details
| Gene Symbol: Gm31087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049679.1 | - | 22 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU848365.1 | - | 1 | 25 | Chr. 18 - 40242753 - 40258495 [-] on Build GRCm38 | |

Gm31087
Genomic Map