siRNA Details
| Gene Symbol: Gm11690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041521.1 | - | 1 | 1395 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB228970.2 | - | 1 | 185 | Chr. 11 - 114219239 - 114237292 [-] on Build GRCm38 | |

Gm11690
Genomic Map