siRNA Details
| Gene Symbol: 9530062E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020618.1 | - | 1 | 479 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722192.1 | - | 1 | 475 | Chr. X - 130393678 - 130394321 [+] on Build GRCm38 | |

9530062E16Rik
Genomic Map