siRNA Details
| Gene Symbol: Cyb561a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040692.1 | - | 238 | |||
| AK139533.1 | - | 1 | 522 | |||
| AK144666.1 | - | 1 | 535 | |||
| AK160179.1 | - | 1 | 238 | |||
| AK168841.1 | - | 1 | 238 | |||
| AK169729.1 | - | 1 | 241 | |||
| AK172638.1 | - | 1 | 241 | |||

Cyb561a3
Genomic Map