siRNA Details
| Gene Symbol: Rsf1os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005667.1 | - | 1 | 48 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705854.1 | - | 1 | 45 | Chr. 7 - 97598349 - 97645112 [-] on Build GRCm38 | |

Rsf1os1
Genomic Map