siRNA Details
| Gene Symbol: Gm30648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050464.1 | - | 394 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB654114.1 | - | 2, 3 | 394 | Chr. 3 - 134200086 - 134211950 [+] on Build GRCm38 | |

Gm30648
Genomic Map