siRNA Details
| Gene Symbol: Gm34753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076685.1 | - | 1 | 137 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB614997.1 | - | 1 | 137 | Chr. 6 - 56369705 - 56371945 [+] on Build GRCm38 | |

Gm34753
Genomic Map