siRNA Details
| Gene Symbol: Gm4466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143754.1 | - | 423 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4466| Gene Symbol: Gm4466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143754.1 | - | 423 | |||