siRNA Details
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037383.1 | - | 2915 | |||
AK087750.1 | - | 2402 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037383.1 | - | 2915 | |||
AK087750.1 | - | 2402 |