siRNA Details
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034423.1 | - | 3 | 3841 | ||
AK037383.1 | - | 2429 | ||||
AK046989.1 | - | 1 | 1481 | |||
AK082754.1 | - | 1 | 1146 | |||
AK087750.1 | - | 1917 |
