siRNA Details
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034423.1 | - | 3 | 2658 | ||
AK035037.1 | - | 1 | 380 | |||
AK037383.1 | - | 1247 | ||||
AK046989.1 | - | 1 | 296 | |||
AK087750.1 | - | 735 |
