siRNA Details
| Gene Symbol: 1110018N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004537.1 | - | 1 | 90 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY704762.1 | - | 1 | 87 | Chr. 2 - 167189109 - 167192494 [+] on Build GRCm38 | |

1110018N20Rik
Genomic Map