siRNA Details
| Gene Symbol: Gm12300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009488.1 | - | 907 | |||
| AV086784.1 | - | 271 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12300| Gene Symbol: Gm12300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009488.1 | - | 907 | |||
| AV086784.1 | - | 271 | ||||