siRNA Details
| Gene Symbol: Gm12300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009488.1 | - | 390 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709201.1 | - | 4 | 388 | Chr. 11 - 67211258 - 67237573 [-] on Build GRCm38 | |

Gm12300
Genomic Map