siRNA Details
Gene Symbol: Gm12300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009488.1 | - | 42 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709201.1 | - | 1 | 42 | Chr. 11 - 67211258 - 67237573 [-] on Build GRCm38 |
