siRNA Details
| Gene Symbol: Gm35439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137696.1 | - | 4 | 555 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722996.1 | - | 4 | 555 | Chr. 3 - 92170221 - 92173736 [+] on Build GRCm38 | |

Gm35439
Genomic Map