siRNA Details
| Gene Symbol: A930009E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035831.1 | - | 1 | 2009 | ||
| BC051426.1 | - | 1 | 3697 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CX215812.1 | - | 1 | 181 | Chr. 6 - 40253561 - 40253959 [+] on Build GRCm38 | |

A930009E05Rik
Genomic Map