siRNA Details
| Gene Symbol: A930009E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020835.1 | - | 1 | 955 | ||
| AK035831.1 | - | 1 | 3102 | |||
| BB125806.2 | - | 1 | 382 | |||
| BB283466.1 | - | 1 | 294 | |||
| BC051426.1 | - | 1 | 4790 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930009E05Rik| Gene Symbol: A930009E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020835.1 | - | 1 | 955 | ||
| AK035831.1 | - | 1 | 3102 | |||
| BB125806.2 | - | 1 | 382 | |||
| BB283466.1 | - | 1 | 294 | |||
| BC051426.1 | - | 1 | 4790 | |||