siRNA Details
| Gene Symbol: 2700038G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012332.1 | - | 1 | 168 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711488.1 | - | 1 | 176 | Chr. 5 - 23853780 - 23854659 [+] on Build GRCm38 | |

2700038G22Rik
Genomic Map