siRNA Details
| Gene Symbol: C330002G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044868.1 | - | 7 | 1790 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C330002G04Rik| Gene Symbol: C330002G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044868.1 | - | 7 | 1790 | ||