siRNA Details
| Gene Symbol: 6430628N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018292.1 | - | 2 | 478 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719165.1 | - | 2 | 475 | Chr. 4 - 115890668 - 115898478 [+] on Build GRCm38 | |

6430628N08Rik
Genomic Map