siRNA Details
| Gene Symbol: 9430030N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020447.1 | - | 1 | 648 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB091595.1 | - | 1 | 203 | Chr. 19 - 45030848 - 45031135 [+] on Build GRCm38 | |

9430030N17Rik
Genomic Map