siRNA Details
| Gene Symbol: Gm15787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040430.1 | 1 | 158 | |||
| NR_040431.1 | 1 | 158 | ||||
| GenBank mRNA | AK053084.1 | - | 1 | 159 | ||
| CJ068547.1 | - | 1 | 121 | |||
| CJ068612.1 | - | 159 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15787| Gene Symbol: Gm15787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040430.1 | 1 | 158 | |||
| NR_040431.1 | 1 | 158 | ||||
| GenBank mRNA | AK053084.1 | - | 1 | 159 | ||
| CJ068547.1 | - | 1 | 121 | |||
| CJ068612.1 | - | 159 | ||||