siRNA Details
| Gene Symbol: Gm15787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040430.1 | 3 | 776 | |||
| NR_040431.1 | 3 | 716 | ||||
| GenBank mRNA | AK053084.1 | - | 3 | 717 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15787| Gene Symbol: Gm15787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040430.1 | 3 | 776 | |||
| NR_040431.1 | 3 | 716 | ||||
| GenBank mRNA | AK053084.1 | - | 3 | 717 | ||