siRNA Details
| Gene Symbol: 1700046C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045918.1 | 4 | 324 | |||
| GenBank mRNA | AK006701.1 | - | 4 | 302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700046C09Rik| Gene Symbol: 1700046C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045918.1 | 4 | 324 | |||
| GenBank mRNA | AK006701.1 | - | 4 | 302 | ||