siRNA Details
Gene Symbol: 1700046C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045918.1 | 4 | 346 | |||
GenBank mRNA | AK006701.1 | - | 4 | 324 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700046C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045918.1 | 4 | 346 | |||
GenBank mRNA | AK006701.1 | - | 4 | 324 |