siRNA Details
| Gene Symbol: Gm10549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045415.1 | 3 | 2536 | |||
| GenBank mRNA | AK141051.1 | - | 3 | 2538 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10549| Gene Symbol: Gm10549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045415.1 | 3 | 2536 | |||
| GenBank mRNA | AK141051.1 | - | 3 | 2538 | ||