siRNA Details
| Gene Symbol: D030045P18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040624.1 | 3 | 1735 | |||
| GenBank mRNA | AK080062.1 | - | 3 | 1736 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D030045P18Rik| Gene Symbol: D030045P18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040624.1 | 3 | 1735 | |||
| GenBank mRNA | AK080062.1 | - | 3 | 1736 | ||