siRNA Details
| Gene Symbol: 9830166K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045314.1 | 2 | 1317 | |||
| GenBank mRNA | AK036719.1 | - | 2 | 1320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9830166K06Rik| Gene Symbol: 9830166K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045314.1 | 2 | 1317 | |||
| GenBank mRNA | AK036719.1 | - | 2 | 1320 | ||