siRNA Details
| Gene Symbol: Gm19424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040320.1 | 3 | 1474 | |||
| GenBank mRNA | AK145978.1 | - | 3 | 1473 | ||
| AK167556.1 | - | 3 | 1473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19424| Gene Symbol: Gm19424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040320.1 | 3 | 1474 | |||
| GenBank mRNA | AK145978.1 | - | 3 | 1473 | ||
| AK167556.1 | - | 3 | 1473 | |||