siRNA Details
| Gene Symbol: Clvs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079058.1 | - | 1494 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clvs2| Gene Symbol: Clvs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079058.1 | - | 1494 | |||