siRNA Details
| Gene Symbol: 1700071M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045444.1 | 3 | 2088 | |||
| NR_045445.1 | 4 | 544 | ||||
| GenBank mRNA | AK133239.1 | - | 3 | 2088 | ||
| AK161361.1 | - | 4 | 545 | |||
| AV274656.2 | - | 2 | 244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700071M16Rik| Gene Symbol: 1700071M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045444.1 | 3 | 2088 | |||
| NR_045445.1 | 4 | 544 | ||||
| GenBank mRNA | AK133239.1 | - | 3 | 2088 | ||
| AK161361.1 | - | 4 | 545 | |||
| AV274656.2 | - | 2 | 244 | |||