siRNA Details
| Gene Symbol: 1700063J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006872.1 | - | 1 | 120 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA465299.1 | - | 1 | 80 | Chr. 17 - 28951851 - 28952343 [-] on Build GRCm38 | |

1700063J08Rik
Genomic Map