siRNA Details
| Gene Symbol: 1700063O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045383.1 | 2 | 166 | |||
| GenBank mRNA | AK006875.1 | - | 2 | 166 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706861.1 | - | 1 | 165 | Chr. 5 - 91766242 - 91767128 [-] on Build GRCm38 | |

1700063O14Rik
Genomic Map