siRNA Details
Gene Symbol: 1700063O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045383.1 | 2 | 50 | |||
GenBank mRNA | AK006875.1 | - | 2 | 50 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706861.1 | - | 1 | 49 | Chr. 5 - 91766242 - 91767128 [-] on Build GRCm38 |
