siRNA Details
Gene Symbol: 1700063O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045383.1 | 1, 2 | 21 | |||
GenBank mRNA | AK006875.1 | - | 1, 2 | 21 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700063O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045383.1 | 1, 2 | 21 | |||
GenBank mRNA | AK006875.1 | - | 1, 2 | 21 |