siRNA Details
| Gene Symbol: Junos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042782.1 | - | 4 | 487 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI079042.1 | - | 4 | 484 | Chr. 4 - 95075669 - 95156278 [+] on Build GRCm38 | |

Junos
Genomic Map